missing translation for 'onlineSavingsMsg'
Learn More

Sodium methoxide, pure, anhydrous powder

Product Code. 10675042 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Unit Size:
100g
2.5kg
500g
5g
This item is not returnable. View return policy

Product Code. 10675042

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 124-41-4 | CH3NaO | 54.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-41-4, 497-19-8, 1310-73-2
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.02
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Alkalinity total alkalinity (as NaOCH3):> 99%
Melting Point 126.0°C
Color White-Yellow
Density 0.8
Packaging Glass bottle
Linear Formula CH3ONa
Quantity 500 g
UN Number 1431
Beilstein 01, 280
Fieser 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
Merck Index 15, 8775
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in methanol and ethanol, insoluble in hydrocarbons(benzene, toluene, xylene
Formula Weight 54.02
Percent Purity ≥97% (NaOCH3)
Grade Pure
Physical Form Powder
Chemical Name or Material Sodium methoxide
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Sodium methoxide, anhydrous powder
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Self-heating substance Category 1
  • Skin corrosion/irritation Category 1 A
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H251-Self-heating: may catch fire.
  • H290-May be corrosive to metals.
  • H228-Flammable solid.
  • EUH014-Reacts violently with water.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235+P410-Keep cool. Protect from sunlight.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado