missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate Dihydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10686652
Click to view available options
Quantity:
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10102-40-6
Molecular Formula MoNa2O4
Molecular Weight (g/mol) 205.93
MDL Number MFCD00149170
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate)
SMILES [Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Color White
pH 9.0 to 10
Physical Form Powder/Solid
Quantity 100 g
Formula Weight 241.95g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'