missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate Dihydrate, puriss., Honeywell Fluka™
BIOPHARMA

Product Code. 15641290
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 2.5 kg

Puriss., meets analytical specification of Ph Eur, 99-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-40-6
Molecular Formula MoNa2O4
Molecular Weight (g/mol) 205.93
MDL Number MFCD00149170
InChI Key TVXXNOYZHKPKGW-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
SMILES [Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Quantity 1 kg
Linear Formula Na2MoO4 · 2H2O
UN Number NONH for all modes of transport
IUPAC Name disodium;dioxido(dioxo)molybdenum;dihydrate
Formula Weight 241.95g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Sodium Molybdate Dihydrate, Meets analytical specification of PhEur
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'