missing translation for 'onlineSavingsMsg'
Learn More

Sodium n-hexadecyl sulfate, 99%, Thermo Scientific Chemicals

Product Code. 11372506
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-hexadecyl sulfate is used as an anionic surfactant.

Solubility
Soluble in water.

Notes
Incompatible with bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-01-0
Molecular Formula C16H33NaO3S
Molecular Weight (g/mol) 328.49
MDL Number MFCD00047766
InChI Key PNGBYKXZVCIZRN-UHFFFAOYSA-M
Synonym sodium hexadecyl sulfate, sodium cetyl sulfate, sodium n-hexadecyl sulfate, unii-3v3y3o7biq, conco sulfate c, avitex c, avitex sf, sodium hexyldecyl sulfate, tergitol anionic 7, 3v3y3o7biq
PubChem CID 23695542
IUPAC Name sodium;hexadecyl sulfate
SMILES [Na+].CCCCCCCCCCCCCCCCS([O-])(=O)=O

Specifications

Melting Point 187°C to 192°C
Quantity 5 g
Beilstein 3578850
Solubility Information Soluble in water.
Formula Weight 344.49
Percent Purity 99%
Chemical Name or Material Sodium n-hexadecyl sulfate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium n-hexadecyl sulfate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.