Learn More
Sodium n-octyl sulfate, 99%, Thermo Scientific Chemicals
Description
Sodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
Melting Point | ∼195°C (decomposition) |
Quantity | 1 g |
Beilstein | 3658635 |
Solubility Information | Soluble in water. |
Formula Weight | 232.28 |
Percent Purity | 99% |
Chemical Name or Material | Sodium n-octyl sulfate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.