Learn More
Sodium nitrate, 99+%, ACS reagent, Thermo Scientific™
Chemical Identifiers
CAS | 7631-99-4 |
---|---|
Molecular Formula | NNaO3 |
Molecular Weight (g/mol) | 84.994 |
InChI Key | VWDWKYIASSYTQR-UHFFFAOYSA-N |
Synonym | sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 |
PubChem CID | 24268 |
ChEBI | CHEBI:63005 |
IUPAC Name | sodium;nitrate |
SMILES | [N+](=O)([O-])[O-].[Na+] |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7631-99-4 | |
84.994 | |
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 | |
CHEBI:63005 | |
[N+](=O)([O-])[O-].[Na+] |
NNaO3 | |
VWDWKYIASSYTQR-UHFFFAOYSA-N | |
24268 | |
sodium;nitrate |
Specifications
7631-99-4 | |
NNaO3 | |
16,86 | |
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1 | |
[N+](=O)([O-])[O-].[Na+] | |
84.994 | |
CHEBI:63005 | |
≥99% | |
Sodium nitrate |
99+% | |
NaNO3 | |
15, 8778 | |
VWDWKYIASSYTQR-UHFFFAOYSA-N | |
sodium;nitrate | |
24268 | |
84.99 | |
ACS Reagent |
Safety and Handling
GHS P Statement May intensify fire; oxidizer. Causes serious eye irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. In case of major fire and large quantities: Evacuate area. Fight fire remotely due to the risk of explosion. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning
- Sodium nitrate
Signal Word
- Warning
Hazard Category
- Oxidising solid Category 3
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
RUO – Research Use Only