Learn More
Sodium nitrite, 99%, extra pure, contains an anticaking reagent, Thermo Scientific Chemicals
64.62 € - 333.00 €
Chemical Identifiers
CAS | 7632-00-0 |
---|---|
Molecular Formula | NNaO2 |
Molecular Weight (g/mol) | 69.00 |
MDL Number | MFCD00011118 |
InChI Key | LPXPTNMVRIOKMN-UHFFFAOYSA-M |
Synonym | sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts |
PubChem CID | 23668193 |
ChEBI | CHEBI:78870 |
IUPAC Name | sodium;nitrite |
SMILES | [Na+].[O-]N=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10742295
|
Thermo Scientific Acros
196620010 |
1 kg | Plastic bottle |
64.62 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10012580
|
Thermo Scientific Acros
196620025 |
2.5 kg | Plastic bottle |
72.46 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10370494
|
Thermo Scientific Acros
196620100 |
10 kg | Plastic Drum |
333.00 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7632-00-0 | |
69.00 | |
LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
23668193 | |
sodium;nitrite |
NNaO2 | |
MFCD00011118 | |
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
CHEBI:78870 | |
[Na+].[O-]N=O |
Specifications
7632-00-0 | |
Crystalline Powder | |
NNaO2 | |
MFCD00011118 | |
15, 8779 | |
LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
sodium;nitrite | |
23668193 | |
69 | |
Extra Pure |
Yellow | |
99% | |
NaNO2 | |
01,1097; 02,386; 04,459; 06,547; 09,432; 11,491; 13,170; 16,86 | |
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
[Na+].[O-]N=O | |
69.00 | |
CHEBI:78870 | |
≥98.5% | |
Sodium nitrite, 99% |
Safety and Handling
GHS P Statement
Toxic if swallowed.
Very toxic to aquatic life.
Causes serious eye irritation.
May intensify fire; oxidizer.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 231-555-9
missing translation for 'rtecsNumber' : RA1225000
missing translation for 'tsca' : TSCA
RUO – Research Use Only