Learn More
Sodium nitrite, ACS, 97% min, Thermo Scientific Chemicals
22.30 € - 57.80 €
Identifiants chimiques
CAS | 7632-00-0 |
---|---|
Molecular Formula | NNaO2 |
Molecular Weight (g/mol) | 69.00 |
MDL Number | MFCD00011118 |
InChI Key | LPXPTNMVRIOKMN-UHFFFAOYSA-M |
Synonym | sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts |
PubChem CID | 23668193 |
ChEBI | CHEBI:78870 |
IUPAC Name | sodium;nitrite |
SMILES | [Na+].[O-]N=O |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
11317817
|
Thermo Scientific Alfa Aesar
014244.14 |
25 g |
22.30 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327817
|
Thermo Scientific Alfa Aesar
014244.22 |
100 g |
30.10 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337817
|
Thermo Scientific Alfa Aesar
014244.36 |
500 g |
57.80 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium nitrite is used in many industrial applications like meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, as an antidote in cyanide poisoning, as an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor as well as in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
7632-00-0 | |
69.00 | |
LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
23668193 | |
sodium;nitrite |
NNaO2 | |
MFCD00011118 | |
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
CHEBI:78870 | |
[Na+].[O-]N=O |
Spécification
271°C | |
Granules | |
NNaO2 | |
sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
sodium;nitrite | |
23668193 | |
69 | |
Hygroscopic | |
Sodium nitrite, For ACS analysis; Decomposes above 320° |
7632-00-0 | |
97% min | |
MFCD00011118 | |
Soluble in water,methanol,ethanol,ammonia and nitric acid. Slightly soluble diethyl ether. | |
[Na+].[O-]N=O | |
69.00 | |
CHEBI:78870 | |
ACS Reagent | |
2.17 g/mL |
Sécurité et traitement
P210-P220-P221-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
H272-H301-H319-H373
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: III
missing translation for 'einecsNumber' : 231-555-9
missing translation for 'rtecsNumber' : RA1225000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only