Learn More
Sodium nitroferricyanide dihydrate, reagent ACS, Thermo Scientific Chemicals
45.02 € - 1394.94 €
Chemical Identifiers
CAS | 13755-38-9 |
---|---|
Molecular Formula | C5H4FeN6Na2O3 |
Molecular Weight (g/mol) | 297.95 |
MDL Number | MFCD00149192 |
InChI Key | JFPDVRGGFBUORC-UHFFFAOYSA-N |
Synonym | nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o |
PubChem CID | 11953895 |
SMILES | O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10239470
|
Thermo Scientific Acros
424360250 |
25 g | Glass bottle |
45.02 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10321651
|
Thermo Scientific Acros
424361000 |
100 g | Glass bottle |
114.42 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10381691
|
Thermo Scientific Acros
424365000 |
500 g | Glass bottle |
364.82 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10107103
|
Thermo Scientific Acros
424360025 |
2.5 kg | Glass bottle |
1394.94 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13755-38-9 | |
297.95 | |
JFPDVRGGFBUORC-UHFFFAOYSA-N | |
11953895 |
C5H4FeN6Na2O3 | |
MFCD00149192 | |
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] |
Specifications
13755-38-9 | |
Crystals | |
Na2Fe(CN)5NO·2H2O | |
15,878 | |
Solubility in water: 400g/L (20°C). Other solubilities: slightly soluble in alcohol | |
O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+] | |
297.95 | |
297.94 | |
ACS Reagent | |
1.7200g/mL |
Bordeaux | |
C5H4FeN6Na2O3 | |
MFCD00149192 | |
nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o | |
JFPDVRGGFBUORC-UHFFFAOYSA-N | |
disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate | |
11953895 | |
99.0 to 102.0% | |
1.72 | |
Sodium nitroferricyanide dihydrate |
Safety and Handling
GHS H Statement
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
missing translation for 'rtecsNumber' : LJ8925000
- Sodium nitroferricyanide dihydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
Hazard Statement
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.