Learn More
Tocris Bioscience™ Sodium orthovanadate
Protein tyrosine phosphatase inhibitor
Brand:  Tocris Bioscience™ 2821/100
101.08 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18742774
  • 129.00 € / 100mg
Explore more special offers
Description
Description
Sodium orthovanadate is an inhibitor of protein tyrosine phosphatases, alkaline phosphatases and ATPases. Suppresses p53-mediated apoptosis by transcription-dependent and -independent pathways.
Specifications
Specifications
100 mg
Sodium orthovanadate
183.91
Sodium orthovanadate