Learn More
Sodium orthovanadate, 99.9% (metals basis), Thermo Scientific Chemicals
inhibitor of Protein Tyrosine Phosphatase and Alkaline Phosphate
93.47 € - 1129.62 €
Chemical Identifiers
CAS | 13721-39-6 |
---|---|
Molecular Formula | Na3O4V |
Molecular Weight (g/mol) | 183.907 |
MDL Number | MFCD00003511 |
InChI Key | IHIXIJGXTJIKRB-UHFFFAOYSA-N |
Synonym | sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt |
PubChem CID | 61671 |
ChEBI | CHEBI:35607 |
IUPAC Name | trisodium;trioxido(oxo)vanadium |
SMILES | [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11384909
|
Thermo Scientific Alfa Aesar
081104.14 |
25 g |
93.47 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11394909
|
Thermo Scientific Alfa Aesar
081104.22 |
100 g |
260.30 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304919
|
Thermo Scientific Alfa Aesar
081104.36 |
500 g |
1129.62 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inhibitor of protein tyrosine phosphatase and alkaline phosphate Sodium orthovanadate is used in protein analysis in molecular biology and acts as an inhibitor of protein tyrosine phosphatases, alkaline phosphatases and kinases. It is also used to prepare the stop solution in dephosphorylation assay of insulin receptor kinase. It serves as a reagent in the development of matrix Chip that utilizes surface-immobilized antibodies. Further, it is utilized for the preservation of protein tyrosyl phosphorylation state in cells and cell lysates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13721-39-6 | |
183.907 | |
IHIXIJGXTJIKRB-UHFFFAOYSA-N | |
61671 | |
trisodium;trioxido(oxo)vanadium |
Na3O4V | |
MFCD00003511 | |
sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt | |
CHEBI:35607 | |
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+] |
Specifications
850°C to 866°C | |
White | |
99.9% (metals basis) | |
MFCD00003511 | |
Soluble in water. | |
[O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+] | |
183.907 | |
CHEBI:35607 | |
Odorless |
13721-39-6 | |
Granules | |
Na3O4V | |
sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt | |
IHIXIJGXTJIKRB-UHFFFAOYSA-N | |
trisodium;trioxido(oxo)vanadium | |
61671 | |
183.94 | |
Sodium orthovanadate |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 237-287-9
missing translation for 'rtecsNumber' : YW1120000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Sodium orthovanadate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Sodium vanadium oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.