Learn More
Sodium oxide, Thermo Scientific Chemicals
100.50 € - 271.35 €
Chemical Identifiers
CAS | 1313-59-3 |
---|---|
Molecular Formula | Na2O |
Molecular Weight (g/mol) | 61.979 |
MDL Number | MFCD00046201 |
InChI Key | KKCBUQHMOMHUOY-UHFFFAOYSA-N |
Synonym | sodium oxide, disodium oxide, disodium monoxide, disodium oxygen-2 anion, disodium oxidandiide, sodium oxide 25g, disodium oxygen 2-, 2na.o |
PubChem CID | 73971 |
IUPAC Name | disodium;oxygen(2-) |
SMILES | [O-2].[Na+].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11304247
|
Thermo Scientific Alfa Aesar
036712.14 |
25 g |
100.50 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314247
|
Thermo Scientific Alfa Aesar
036712.22 |
100 g |
271.35 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium oxide is used in chemical manufacturing, ceramics and glasses. Ethylbenzene dehydrogenation and carbon dioxide shift-reaction occurs using a sodium oxide/alumina catalyst. It is employed as catalyzing gasification of CO2 on carbon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-59-3 | |
61.979 | |
KKCBUQHMOMHUOY-UHFFFAOYSA-N | |
73971 | |
[O-2].[Na+].[Na+] |
Na2O | |
MFCD00046201 | |
sodium oxide, disodium oxide, disodium monoxide, disodium oxygen-2 anion, disodium oxidandiide, sodium oxide 25g, disodium oxygen 2-, 2na.o | |
disodium;oxygen(2-) |
Specifications
>400°C (decomposition) | |
Beads | |
MFCD00046201 | |
14,8651 | |
Soluble in water. | |
[O-2].[Na+].[Na+] | |
61.979 | |
61.98 | |
2.27 g/mL |
1313-59-3 | |
Na2O | |
UN1825 | |
sodium oxide, disodium oxide, disodium monoxide, disodium oxygen-2 anion, disodium oxidandiide, sodium oxide 25g, disodium oxygen 2-, 2na.o | |
KKCBUQHMOMHUOY-UHFFFAOYSA-N | |
disodium;oxygen(2-) | |
73971 | |
Air and moisture sensitive | |
Sodium oxide |
Safety and Handling
GHS H Statement
H271-H314
P210-P220-P221-P260-P264b-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P501c
H271-H314-H335
missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: I
missing translation for 'einecsNumber' : 215-208-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.