missing translation for 'onlineSavingsMsg'
Learn More

Sodium perchlorate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10752241
Click to view available options
Quantity:
5 g
250 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-89-0
InChI Key BAZAXWOYCMUHIX-UHFFFAOYSA-M
Synonym sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
PubChem CID 522606
IUPAC Name sodium;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Na+]

Specifications

Melting Point 482.0°C
Color White
pH 6.0 to 8.0 (5% aq. soln.)
Physical Form Crystalline Powder and/or Crystals
Quantity 250 g
Assay Percent Range 99% min. (on dry substance)
Molecular Formula ClNaO4
Linear Formula NaClO4
Merck Index 15,8784
Solubility Information Solubility in water: 209 g/100mL (15°C). Other solubilities: 170 g/100mL water (0°C), 300 g/100mL water (80°C), soluble in alcohol
Molecular Weight (g/mol) 122.44
Formula Weight 122.44
Percent Purity ≥99%
Grade Analytical
Loss on Drying 2.0% max. (130°C, 2 hrs)
Packaging Plastic bottle
Chemical Name or Material Sodium perchlorate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium perchlorate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.