missing translation for 'onlineSavingsMsg'
Learn More

Sodium perchlorate, anhydrous, ACS, 98.0-102.0%, Thermo Scientific Chemicals

49.14 € - 145.73 €

Chemical Identifiers

CAS 7601-89-0
Molecular Formula ClNaO4
Molecular Weight (g/mol) 122.436
MDL Number MFCD00003480
InChI Key BAZAXWOYCMUHIX-UHFFFAOYSA-M
Synonym sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
PubChem CID 522606
IUPAC Name sodium;perchlorate
SMILES [O-]Cl(=O)(=O)=O.[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381718
View Documents
Thermo Scientific Alfa Aesar
011623.22
100 g
49.14 €
100g
Please to purchase this item. Need a web account? Register with us today!
11371718
View Documents
Thermo Scientific Alfa Aesar
011623.36
500 g
145.73 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium perchlorate is used in chemical analysis and in explosive materials, fireworks, and rocket motors. It is the precursor for the preparation of other perchlorate salts. It is also employed as a chaotrophic agent in DNA extraction and hybridization reactions in molecular biology. It prevents the binding of phenolic compounds to RNA during extraction. In addition, it is used in batteries, plastics and in rubber products. It is also used as a nonreactive electrolyte. Further, it is an eluent component used for analysis of biological samples by liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7601-89-0
122.436
BAZAXWOYCMUHIX-UHFFFAOYSA-M
522606
[O-]Cl(=O)(=O)=O.[Na+]
ClNaO4
MFCD00003480
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
sodium;perchlorate

Specifications

482°C (decomposition)
Crystalline
ClNaO4
sodium perchlorate, perchloric acid, sodium salt, natriumperchlorat, perchlorate de sodium, natriumperchloraat dutch, natriumperchlorat german, unii-97f4mty3va, cpl 46, perchlorate de sodium french, sodio perclorato di italian
BAZAXWOYCMUHIX-UHFFFAOYSA-M
sodium;perchlorate
522606
ACS Reagent
2.50 g/mL
7601-89-0
98.0 to 102.0%
MFCD00003480
Soluble in water and alcohol.
[O-]Cl(=O)(=O)=O.[Na+]
122.436
122.46
Hygroscopic
Sodium perchlorate, anhydrous, For analysis ACS
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P260-P264b-P270-P280-P283-P301+P312-P305+P351+P338-P306+P360-P314-P330-P370+P378q-P371+P380+P375-P501c

H271-H302-H319-H373

missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II

missing translation for 'einecsNumber' : 231-511-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Sodium perchlorate, anhydrous

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.

Supplemental information
  • MIXTURE LIST-Contains : Sodium perchlorate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only