missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Perchlorate Monohydrate,≥99% for HPLC, Honeywell Fluka™
for HPLC,≥99.0%
63.77 € - 246.27 €
Chemical Identifiers
CAS | 7791-07-3 |
---|---|
Molecular Formula | ClH2NaO5 |
Molecular Weight (g/mol) | 140.45 |
MDL Number | MFCD00149164 |
InChI Key | IXGNPUSUVRTQGW-UHFFFAOYSA-M |
Synonym | sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate |
PubChem CID | 516899 |
IUPAC Name | sodium;perchlorate;hydrate |
SMILES | O.[O-]Cl(=O)(=O)=O.[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15687380
|
Honeywell-Fluka
89152-50G |
50 g |
63.77 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15677380
|
Honeywell-Fluka
89152-250G |
250 g |
246.27 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7791-07-3 | |
140.45 | |
IXGNPUSUVRTQGW-UHFFFAOYSA-M | |
516899 | |
O.[O-]Cl(=O)(=O)=O.[Na+] |
ClH2NaO5 | |
MFCD00149164 | |
sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate | |
sodium;perchlorate;hydrate |
Specifications
7791-07-3 | |
NaClO4 · H2O | |
UN1502 | |
IXGNPUSUVRTQGW-UHFFFAOYSA-M | |
sodium;perchlorate;hydrate | |
516899 | |
≥99% (T), ≥99% |
ClH2NaO5 | |
MFCD00149164 | |
sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate | |
O.[O-]Cl(=O)(=O)=O.[Na+] | |
140.45 | |
140.46g/mol | |
Sodium perchlorate monohydrate |
Safety and Handling
P220
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Sodium perchlorate monohydrate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 1
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H271-May cause fire or explosion; strong oxidiser.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.