missing translation for 'onlineSavingsMsg'
Learn More

Sodium periodate, 99.8+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10229990
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Melting Point 300.0°C
Color White
Physical Form Crystalline Powder
Quantity 500 g
Assay Percent Range 99.8 to 100.3% (on dried basis) (Iodometry)
Linear Formula NaIO4
Fieser 01,809; 02,311; 04,373; 05,507; 06,455; 11,492; 13,145; 15,294
Merck Index 15,8772
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: 389g/L (52°C), soluble in h2so4, hno3, acetic acids, insoluble in most common organic solvents
IUPAC Name sodium;periodate
Formula Weight 213.89
Percent Purity ≥99.8%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Sodium periodate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium periodate
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'