missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium periodate, 99.8+%, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Specifications
Melting Point | 300.0°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 100 g |
Assay Percent Range | 99.8 to 100.3% (on dried basis) (Iodometry) |
Linear Formula | NaIO4 |
Fieser | 01,809; 02,311; 04,373; 05,507; 06,455; 11,492; 13,145; 15,294 |
Merck Index | 15,8772 |
Solubility Information | Solubility in water: 80g/L (20°C). Other solubilities: 389g/L (52°C),soluble in h2so4,hno3,acetic acids,insoluble in most common organic solvents |
IUPAC Name | sodium;periodate |
Show More |
Safety and Handling
Product Identifier
- Sodium periodate
- Danger
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Oxidising solid Category 1
- Skin corrosion/irritation Category 1 C
- Specific target organ toxicity Category 1
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H271-May cause fire or explosion; strong oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'