missing translation for 'onlineSavingsMsg'
Learn More

Sodium Periodate, Certified AR for Analysis, Fisher Chemical™

Product Code. 11413753
Click to view available options
Quantity:
100 g
250 g
5 kg

INaO4, CAS Number-7790-28-5, sodiumperiodate, sodium m-periodate, periodate sodium, sodium periodate, periodic acid hio4 , sodium salt, sodium meta periodate, sodium penodate, sodium metaperiodate, periodic acid, sodium salt, sodium meta-periodate, 250g, CHEBI:75226, White, 213.89g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
IUPAC Name sodium;periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Melting Point 300°C
Color White
pH 3.4 to 4.5
Physical Form Solid
Quantity 5 kg
Formula Weight 213.89g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium periodate
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 C
  • Specific target organ toxicity Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P221-Take any precaution to avoid mixing with combustibles A§.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'