missing translation for 'onlineSavingsMsg'
Learn More

Sodium persulfate, +98%, Thermo Scientific Chemicals

Product Code. 10318580
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7775-27-1
Molecular Formula Na2O8S2
Molecular Weight (g/mol) 238.092
MDL Number MFCD00003501
InChI Key CHQMHPLRPQMAMX-UHFFFAOYSA-L
Synonym sodium persulfate, sodium peroxydisulfate, sodium peroxodisulfate, peroxydisulfuric acid, disodium salt, disodium peroxodisulphate, sodium persulphate, unii-j49fyf16je, persulfate de sodium french, sodium peroxydisulphate, disodium sulfonatooxy sulfate
PubChem CID 62655
IUPAC Name disodium;sulfonatooxy sulfate
SMILES [O-]S(=O)(=O)OOS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White to Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 98+%
Linear Formula Na2S2O8
Fieser 01,1102; 03,267
Merck Index 15, 8787
Formula Weight 238.09
Percent Purity ≥98.0% (Iodometry)
Identification Pass Test
Packaging Plastic bottle
Chemical Name or Material Sodium persulfate, 98+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Sodium Persulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.