missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Persulfate, Honeywell
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Quantity | 2.5 kg |
Linear Formula | Na2S2O8 |
UN Number | 1505 |
Formula Weight | 238.1g/mol |
Percent Purity | ≥98% |
Grade | Reagent |
Chemical Name or Material | Sodium Persulfate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction