Learn More
Thermo Scientific Chemicals Sodium phosphate, 0.2M buffer soln., pH 7.4
85.02 € - 119.60 €
Specifications
Chemical Name or Material | Sodium phosphate buffer soln. |
---|---|
Concentration | 0.2 M |
pH | 7.4 |
Physical Form | Liquid |
MDL Number | MFCD00146206 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15494569
|
Thermo Scientific Chemicals
J62152.AP |
500 mL |
85.02 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15404579
|
Thermo Scientific Chemicals
J62152.K2 |
1 L |
119.60 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately. It is also used in the preparation of mobile phase in high performance liquid chromatography (HPLC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBuffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately. It is also used in the preparation of mobile phase in high performance liquid chromatography (HPLC).
Solubility
Miscible with water. Immiscible with alcohol.
Notes
Incompatible with strong acids.
Specifications
Sodium phosphate buffer soln. | |
7.4 | |
MFCD00146206 |
0.2 M | |
Liquid | |
Miscible with water. Immiscible with alcohol. |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.