Learn More
Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 7.6
Brand:  Thermo Scientific Chemicals J60158.AK
83.10 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15432979
  • 83.52 € / 250mL
Explore more special offers
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sodium phosphate buffer soln.
7.6
250 mL
0.5 M
Liquid
MFCD00146206
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 7.6 > 250mL