missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 7.6

Brand:  Thermo Scientific Chemicals J60158.AP

132.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 15442979

  • 132.66 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sodium phosphate buffer soln.
7.6
500 mL
0.5 M
Liquid
MFCD00146206
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ28K509 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberJ60158.AP,J60158.AK
Lot NumberM08K518 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog NumberJ60158.AP,J60158.AK
Lot NumberW28J516 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog NumberJ60158.AP,J60158.AK
Lot NumberR02J540 Certificate TypeCertificate of Analysis Date26/06/2023 Catalog NumberJ60158,J60158.AP,J60158.AK
Lot NumberR02J539 Certificate TypeCertificate of Analysis Date26/06/2023 Catalog NumberJ60158,J60158.AP,J60158.AK
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 7.6 > 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only