Learn More
Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 9.5
78.80 € - 125.00 €
Specifications
Chemical Name or Material | Sodium phosphate buffer soln. |
---|---|
Concentration | 0.5 M |
pH | 9.5 |
Physical Form | Liquid |
MDL Number | MFCD00146206 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15484049
|
Thermo Scientific Chemicals
J61456.AK |
250 mL |
78.80 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15494049
|
Thermo Scientific Chemicals
J61456.AP |
500 mL |
125.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium phosphate buffer soln. | |
9.5 | |
MFCD00146206 |
0.5 M | |
Liquid |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.