missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic, 98+%, extra pure, anhydrous, Thermo Scientific Chemicals

50.35 € - 705.51 €

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12665147
View Documents
Thermo Scientific Acros
448140010
1 kg
50.35 €
1kg
Please to purchase this item. Need a web account? Register with us today!
12675147
View Documents
Thermo Scientific Acros
448140050
5 kg
179.90 €
5kg
Please to purchase this item. Need a web account? Register with us today!
12685147
View Documents
Thermo Scientific Acros
448140250
25 kg
705.51 €
25kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7558-79-4
141.96
BNIILDVGGAEEIG-UHFFFAOYSA-L
24203
[Na+].[Na+].OP([O-])([O-])=O
HNa2O4P
MFCD00003496
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
CHEBI:34683

Specifications

7558-79-4
HNa2O4P
MFCD00003496
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium;hydrogen phosphate
24203
141.96
Extra Pure
Sodium phosphate, dibasic
98+%
Na2HPO4
15,8789
Solubility in water: 75g/L (20°C). Other solubilities: practically insoluble in alcohol
[Na+].[Na+].OP([O-])([O-])=O
141.96
CHEBI:34683
≥98%
Plastic bottle
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-448-7

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.