missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic, ≥99.999% (metals basis), Honeywell™ Fluka™ TraceSELECT™

Product Code. 15666800 Shop All Honeywell Fluka Products
100 g, Poly bottle
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

TraceSELECT™, for trace analysis, anhydrous,≥99.999% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions

 


 

TRUSTED_SUSTAINABILITY

Specifications

CAS 7558-79-4
Quantity 100 g
Molecular Formula HNa2O4P
Linear Formula Na2HPO4
MDL Number MFCD00003496
UN Number UN3077
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
SMILES [Na+].[Na+].OP([O-])([O-])=O
IUPAC Name disodium;hydrogen phosphate
Molecular Weight (g/mol) 141.96
PubChem CID 24203
ChEBI CHEBI:34683
Formula Weight 141.96g/mol
Percent Purity ≥99.999% (metals basis)
Packaging Poly bottle
Chemical Name or Material Sodium phosphate dibasic
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Phosphate Dibasic, ≥99.999% (metals basis), Honeywell™ Fluka™ TraceSELECT™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.