missing translation for 'onlineSavingsMsg'
Learn More
Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions, Thermo Scientific Chemicals Product Code.: 10782602

Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions, Thermo Scientific Chemicals

Product Code. 10782602
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10782602

Brand: Thermo Scientific Acros 424375000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

pH buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
pH 8.7 to 9.3 (5% soln. at 25°C)
Physical Form Granular Powder
Quantity 500 g
Assay Percent Range 99+%
Linear Formula Na2HPO4
Merck Index 15, 8789
IUPAC Name disodium hydrogen phosphate
Formula Weight 141.96
Percent Purity ≥99%
Grade ACS Reagent
Loss on Drying 0.2% max. (105°C)
Packaging Plastic bottle
Decomposition Information Decomposes information: > 240°C
Chemical Name or Material Sodium phosphate, dibasic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium phosphate, dibasic, 99+%, ACS reagent, suitable for buffer solutions, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.