missing translation for 'onlineSavingsMsg'
Learn More

Sodium phosphate, dibasic, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

23.52 € - 241.20 €

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium saltShow MoreShow Less
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10361474
SDS View Documents
Thermo Scientific Acros
204851000
100 g Plastic bottle
23.52 €
100g
Please to purchase this item. Need a web account? Register with us today!
10440481
SDS View Documents
Thermo Scientific Acros
204855000
500 g Plastic bottle
49.99 €
500g
Please to purchase this item. Need a web account? Register with us today!
10698504
SDS View Documents
Thermo Scientific Acros
204850050
5 kg Plastic Bottle
241.20 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer for analysis
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

missing translation for 'einecsNumber' : 231-448-7

missing translation for 'rtecsNumber' : WC4500000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium phosphate, dibasic, 99+%, for analysis, anhydrous, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.