missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Phosphate, Dibasic, Anhydrous, Powder, Ultrapure Bioreagent, J.T. Baker™
High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography
244.00 € - 552.00 €
Chemical Identifiers
CAS | 7558-79-4 |
---|---|
Molecular Formula | HNa2O4P |
Molecular Weight (g/mol) | 141.96 |
MDL Number | MFCD00003496 |
InChI Key | BNIILDVGGAEEIG-UHFFFAOYSA-L |
Synonym | disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt |
PubChem CID | 24203 |
ChEBI | CHEBI:34683 |
IUPAC Name | disodium;hydrogen phosphate |
SMILES | [Na+].[Na+].OP([O-])([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13285423
|
Avantor J.T.Baker
4062-01 |
500 g | Poly Bottle |
244.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15326175
|
Avantor J.T.Baker
4062-05 |
2.5 kg | Poly Bottle |
552.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For Liquid Chromatography and Molecular Biology applications
Chemical Identifiers
7558-79-4 | |
141.96 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
24203 | |
disodium;hydrogen phosphate |
HNa2O4P | |
MFCD00003496 | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
CHEBI:34683 | |
[Na+].[Na+].OP([O-])([O-])=O |
Specifications
7558-79-4 | |
HNa2O4P | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
[Na+].[Na+].OP([O-])([O-])=O | |
141.96 | |
CHEBI:34683 | |
Ultrapure Bioreagent | |
Sodium Phosphate, Dibasic, Anhydrous |
Powder | |
MFCD00003496 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
disodium;hydrogen phosphate | |
24203 | |
141.96 | |
1L = 2.07kg |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title