missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic dihydrate, meets analytical spec. of BP, Ph. Eur., 98.5-101% (calc. to the dried substance), Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
38.39 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 kg |
Linear Formula | Na2HPO4 · 2H2O |
IUPAC Name | disodium;hydrogen phosphate;dihydrate |
Formula Weight | 177.99g/mol |
Percent Purity | 98.5 to 101% (calc. to the dried substance) |
Chemical Name or Material | Sodium Phosphate Dibasic Dihydrate, Meets analytical specification of Ph.Eur.BP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'