missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic Dihydrate, puriss. p.a., Honeywell Fluka™

Product Code. p-7071969 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
6 x 1 kg
Unit Size:
1kg
500g
5kg
Pack of 6
This item is not returnable. View return policy

Product Code. 15613040

Brand: Honeywell-Fluka 30435500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., Reagent European Pharmacopoeia, 98.5-101.0% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
PubChem CID 6451167
IUPAC Name disodium;hydrogen phosphate;dihydrate
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 500 g
Linear Formula Na2HPO4 · 2H2O
Formula Weight 177.99g/mol
Percent Purity 98.5 to 101% (calc. to the dried substance)
Grade puriss. p.a.
Chemical Name or Material Sodium Phosphate Dibasic Dihydrate, Reag. Ph. Eur.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.