missing translation for 'onlineSavingsMsg'
Learn More

Sodium Phosphate Dibasic, MP Biomedicals™

Product Code. 11454652
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Widely used in molecular biology, analytical chemistry, biochemistry and chromatography. Sodium Phosphate Dibasic, MP Biomedicals is a reagent with very high buffering capacity, often used in conjunction with Sodium Phosphate, Monobasic in the preparation

Sodium Phosphate Dibasic is used in many applications, including the purification of antibodies, as a reagent and buffer in analytical chemistry, and in the preparation of biological buffers. Also used as a sequestrant, emulsifier and buffer in foods; as mordant in dyeing; for weighting silk; in tanning; in manufacturing of enamels, ceramics, detergents, and boiler compounds; as a fireproofing agent in soldering and brazing instead of borax; as a cathartic; and as a laxative.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Solubility Information Soluble in water, but more soluble in hot water; insoluble in alcohol, ethanol.
IUPAC Name disodium hydrogen phosphate
Formula Weight 141.96
Percent Purity ≥99.0%
Grade ACS
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.