missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate dibasic, Puriss., meets analytical spec. of BP, Ph. Eur., USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance), Honeywell Fluka™
Click to view available options
Quantity:
1 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Physical Form | Anhydrous |
Quantity | 25 kg |
Linear Formula | Na2HPO4 |
Solubility Information | Very Soluble in hot water, 8 parts soluble in water , insoluble in alcohol |
Formula Weight | 141.96g/mol |
Percent Purity | 98 to 100.5% (calc. to the dried substance) |
Grade | puriss. |
Chemical Name or Material | Sodium Phosphate Dibasic, Meets analytical specification of Ph.Eur.BP, USP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'