missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate, monobasic, 99%, for analysis, anhydrous, Thermo Scientific Chemicals
29.25 € - 522.60 €
Chemical Identifiers
CAS | 7558-80-7 |
---|---|
Molecular Formula | H2NaO4P |
Molecular Weight (g/mol) | 119.98 |
MDL Number | MFCD00003527,MFCD00146206 |
InChI Key | AJPJDKMHJJGVTQ-UHFFFAOYSA-M |
Synonym | monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate |
PubChem CID | 23672064 |
ChEBI | CHEBI:37585 |
SMILES | [Na+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10751135
|
Thermo Scientific Acros
389872500 |
250 g | Plastic bottle |
29.25 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10135013
|
Thermo Scientific Acros
389870010 |
1 kg | Plastic bottle |
75.58 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11350836
|
Thermo Scientific Acros
389870025 |
2.5 kg | Plastic bottle |
157.79 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914291
|
Thermo Scientific Acros
389870100 |
10 kg | Plastic drum |
522.60 €
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological bufferChemical Identifiers
7558-80-7 | |
119.98 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
23672064 | |
[Na+].OP(O)([O-])=O |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
CHEBI:37585 |
Specifications
7558-80-7 | |
4.1 to 4.7 (5% soln.) | |
99% | |
NaH2PO4 | |
15,879 | |
Solubility in water: 850g/L (20°C). | |
[Na+].OP(O)([O-])=O | |
119.98 | |
CHEBI:37585 | |
99% | |
Sodium phosphate, monobasic |
White | |
Crystalline Powder | |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
sodium;dihydrogen phosphate | |
23672064 | |
119.98 | |
Analytical |
Safety and Handling
missing translation for 'einecsNumber' : 231-449-2
Product Identifier
- Sodium phosphate, monobasic
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title