missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate, monobasic dihydrate, 98%, extra pure, Thermo Scientific Chemicals
46.70 € - 416.00 €
Chemical Identifiers
CAS | 13472-35-0 |
---|---|
Molecular Formula | H2NaO4P·2H2O |
Molecular Weight (g/mol) | 156.01 |
InChI Key | VBJGJHBYWREJQD-UHFFFAOYSA-M |
Synonym | sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh |
PubChem CID | 23673460 |
IUPAC Name | sodium;dihydrogen phosphate;dihydrate |
SMILES | O.O.OP(=O)(O)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12645107
|
Thermo Scientific Acros
447760010 |
1 kg |
46.70 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12655107
|
Thermo Scientific Acros
447760050 |
5 kg |
141.00 €
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12665107
|
Thermo Scientific Acros
447760250 |
25 kg |
416.00 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13472-35-0 | |
156.01 | |
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
sodium;dihydrogen phosphate;dihydrate |
H2NaO4P·2H2O | |
VBJGJHBYWREJQD-UHFFFAOYSA-M | |
23673460 | |
O.O.OP(=O)(O)[O-].[Na+] |
Specifications
60.0°C | |
98% | |
NaH2PO4·2H2O | |
sodium dihydrogen phosphate dihydrate, sodium phosphate monobasic dihydrate, monosodium phosphate dihydrate, sodium phosphate, monobasic, dihydrate, phosphoric acid, monosodium salt, dihydrate, sodium dihydrogenphosphate dihydrate, sodium phosphate nah2po4 dihydrate, monosodium dihydrogen phosphate dihydrate, sodium dihydrate dihydrogen phosphate, acmc-20ajoh | |
VBJGJHBYWREJQD-UHFFFAOYSA-M | |
sodium;dihydrogen phosphate;dihydrate | |
23673460 | |
98% | |
Plastic bottle |
13472-35-0 | |
H2NaO4P·2H2O | |
15,879 | |
Solubility in water: soluble. Other solubilities: practically insoluble in alcohol | |
O.O.OP(=O)(O)[O-].[Na+] | |
156.01 | |
156.01 | |
Extra Pure | |
Sodium phosphate, monobasic dihydrate |
RUO – Research Use Only