missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate monobasic dihydrate, meets analytical spec. of BP, FCC, Ph. Eur., USP, E 339, 98-100.5% (calc. to the dried substance), Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
53.59 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 kg |
Linear Formula | NaH2PO4 · 2H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 156.01g/mol |
Percent Purity | 98 to 100.5% (calc. to the dried substance) |
Chemical Name or Material | Sodium Phosphate Monobasic Dihydrate, Meets analytical specification of Ph. Eur., BP, USP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'