missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate monobasic, For HPLC, ≥99.0%, Honeywell Fluka™
for HPLC,≥99.0%
118.20 € - 458.93 €
Chemical Identifiers
CAS | 7558-80-7 |
---|---|
Molecular Formula | H2NaO4P |
Molecular Weight (g/mol) | 119.98 |
MDL Number | MFCD00003527,MFCD00146206 |
InChI Key | AJPJDKMHJJGVTQ-UHFFFAOYSA-M |
Synonym | monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate |
PubChem CID | 23672064 |
ChEBI | CHEBI:37585 |
IUPAC Name | sodium;dihydrogen phosphate |
SMILES | [Na+].OP(O)([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15601590
|
Honeywell-Fluka
17844-50G |
50 g |
118.20 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15691580
|
Honeywell-Fluka
17844-250G |
250 g |
458.93 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
7558-80-7 | |
119.98 | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
23672064 | |
sodium;dihydrogen phosphate |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
CHEBI:37585 | |
[Na+].OP(O)([O-])=O |
Specifications
7558-80-7 | |
H2NaO4P | |
MFCD00003527,MFCD00146206 | |
monosodium phosphate, sodium dihydrogen phosphate, sodium dihydrogenorthophosphate, sodium phosphate monobasic, sodium acid phosphate, sodium phosphate, monobasic, monosodium dihydrogen orthophosphate, sodium phosphate monobasic anhydrous, monosodium orthophosphate, monobasic sodium phosphate | |
AJPJDKMHJJGVTQ-UHFFFAOYSA-M | |
sodium;dihydrogen phosphate | |
23672064 | |
119.98 | |
HPLC | |
2.36 g/cm3 |
100.0000% | |
NaH2PO4 | |
NONH for all modes of transport | |
Soluble in water 0.1g/mL at 25°C | |
[Na+].OP(O)([O-])=O | |
119.98 | |
CHEBI:37585 | |
≥99% | |
Glass Bottle | |
Sodium phosphate monobasic, For HPLC, ≥99.0% |
Safety and Handling
missing translation for 'shelfLife' : 1620 days from date of manufacture
missing translation for 'einecsNumber' : 231-449-2
missing translation for 'rtecsNumber' : WA1900000
Recommended Storage : Room Temp