missing translation for 'onlineSavingsMsg'
Learn More
Sodium phosphate, monobasic monohydrate, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10513544

Sodium phosphate, monobasic monohydrate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10513544
250 g, Plastic bottle
Click to view available options
Quantity:
250 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10513544

Brand: Thermo Scientific Acros 207802500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
Synonym sodium dihydrogen phosphate monohydrate, sodium phosphate monobasic monohydrate, monosodium phosphate monohydrate, unii-593yog76rn, phosphoric acid, monosodium salt, monohydrate, sodium phosphate monobasic hydrate, sodium dihydrogen phosphate hydrate, sodium dihydrogenphosphate monohydrate, sodium hydrate dihydrogen phosphate, pubchem12707Show MoreShow Less
PubChem CID 516949
SMILES O.[Na+].OP(O)(O)=O

Specifications

Melting Point 100.0°C
Color White
pH 4.1 to 4.5 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 250 g
Assay Percent Range 99+%
Linear Formula NaH2PO4·H2O
Fieser 01,395
Merck Index 15, 8790
Solubility Information Solubility in water: 1103g/L (20°C). Other solubilities: practically insoluble in alcohol
IUPAC Name sodium;dihydrogen phosphate;hydrate
Formula Weight 137.99
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Sodium phosphate, monobasic monohydrate, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462399 Certificate TypeCertificate of Analysis Date01/05/2024 Catalog Number207800025,207805000,207802500
Lot NumberA0454032 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number207800025,207805000,207802500
Lot NumberA0399326 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number207800025,207805000,207802500
Lot NumberA0448359 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number207800025,207805000,207802500
Lot NumberA0406347 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number207800025,207805000,207802500
Product Identifier
  • Sodium phosphate, monobasic monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium phosphate, monobasic monohydrate, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.