missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium phosphate, tribasic dodecahydrate, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 75.0°C |
Quantity | 1 kg |
Linear Formula | Na3PO4·12H2O |
Merck Index | 15,8792 |
Solubility Information | Solubility in water: 240g/L (20°C). Other solubilities: practically insoluble in alcohol |
Formula Weight | 380.13 |
Grade | Extra Pure |
Packaging | Plastic bottle |
Specific Gravity | 1.62 |
Density | 1.6200g/mL |
Show More |
Safety and Handling
Product Identifier
- Sodium phosphate, tribasic dodecahydrate
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'