missing translation for 'onlineSavingsMsg'
Learn More

Sodium pyruvate, 99%

Product Code. 11439063
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
This item is not returnable. View return policy

Product Code. 11439063

Brand: Thermo Scientific Alfa Aesar A11148.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Sodium pyruvate, 99%, C3H3NaO3, CAS Number-113-24-6, 2-oxopropanoic acid sodium salt, pod38aif08, propanoic acid, 2-oxo-, sodium salt, pyruvate sodium, pyruvic acid sodium salt, pyruvic acid, sodium salt, sodium 2-oxopropanoate, sodium alpha-ketopropionate, sodium pyruvate, unii-pod38aif08, 500g | CAS: 113-24-6 | C3H3NaO3 | 110.044 g/mol

Sodium pyruvate is used in cell culture media as an additional source of energy. It acts as an antioxidant and finds protective effects against oxygen radicals. It serves as an intermediate in many metabolic pathways such as sugar metabolism. It is converted into acetyl coenzyme A and enters the trichloroacetic acid cycle. In addition, it is involved with amino acid metabolism and initiates the Kreb's cycle. It plays an important role as a free radical scavenger.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium pyruvate is used in cell culture media as an additional source of energy. It acts as an antioxidant and finds protective effects against oxygen radicals. It serves as an intermediate in many metabolic pathways such as sugar metabolism. It is converted into acetyl coenzyme A and enters the trichloroacetic acid cycle. In addition, it is involved with amino acid metabolism and initiates the Kreb′s cycle. It plays an important role as a free radical scavenger.

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.044
MDL Number MFCD00002586
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
PubChem CID 23662274
ChEBI CHEBI:50144
IUPAC Name sodium;2-oxopropanoate
SMILES CC(=O)C(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 50 g
Beilstein 3568341
Solubility Information Soluble in water.
Formula Weight 110.04
Percent Purity 99%
Chemical Name or Material Sodium pyruvate
compliance-icons
Nombre del producto
  • Sodium pyruvate
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Indicaciones de peligro
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contains : Sodium pyruvate

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado