missing translation for 'onlineSavingsMsg'
Learn More
Sodium selenite, 44-46% Se, anhydrous, Thermo Scientific Chemicals Product Code.: 10701481

Sodium selenite, 44-46% Se, anhydrous, Thermo Scientific Chemicals

Product Code. 10701481
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10701481

Brand: Thermo Scientific Acros 200731000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-18-8
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.94
MDL Number MFCD00003489
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
Synonym sodium selenite, disodium selenite, natriumselenit, selenious acid, disodium salt, natriumselenit german, disodium selenium trioxide, unii-hiw548rq3w, selenious acid disodium salt, ccris 1260, hsdb 768
PubChem CID 24934
ChEBI CHEBI:48843
IUPAC Name disodium;selenite
SMILES [O-][Se](=O)[O-].[Na+].[Na+]

Specifications

Melting Point 350°C
Color White to Beige
Physical Form Powder
Quantity 100 g
Linear Formula Na2SeO3
Merck Index 15, 8802
Solubility Information Solubility in water: 950g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 172.94
Percent Purity 44 to 46% Se
Loss on Drying 0.5% max.
Packaging Glass bottle
Chemical Name or Material Sodium selenite, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463726 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number200730250,200735000,200731000
Lot NumberA0412523 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number200730250,200735000,200731000
Lot NumberA0446714 Certificate TypeCertificate of Analysis Date20/10/2022 Catalog Number200730250,200735000,200731000
Lot NumberA0416979 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog Number200730250,200735000,200731000
compliance-icons compliance-icons
Product Identifier
  • Sodium selenite
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium selenite, 44-46% Se, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.