missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Selenite Pentahydrate, Honeywell Fluka™
Click to view available options
Quantity:
25 g
100 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 100 g |
Linear Formula | Na2SeO3 · 5H2O |
UN Number | 2630 |
Formula Weight | 263.01g/mol |
Percent Purity | ≥99% (RT) |
Chemical Name or Material | Sodium Selenite Pentahydrate |
Safety and Handling
Product Identifier
- Sodium selenite pentahydrate
- Danger
- Acute toxicity Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- H300 + H330-Fatal if swallowed or if inhaled.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'