missing translation for 'onlineSavingsMsg'
Learn More

Sodium Selenite Pentahydrate, Honeywell Fluka™

Product Code. 15670510
Click to view available options
Quantity:
25 g
100 g

Puriss. p.a.,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26970-82-1
Molecular Formula H10Na2O8Se
Molecular Weight (g/mol) 263.023
MDL Number MFCD00149173
InChI Key TUANAMBRHOLYTH-UHFFFAOYSA-L
Synonym sodium selenite pentahydrate, disodium selenite pentahydrate, unii-0wv4l961zv, ccris 1378, na2seo3.5h2o, selenious acid, disodium salt, pentahydrate, disodium pentahydrate selenite, sodium selenite hydrate 2:1:5, sodium selenite hydrate, disodium selenite hydrate
PubChem CID 134930
IUPAC Name disodium;selenite;pentahydrate
SMILES O.O.O.O.O.[O-][Se](=O)[O-].[Na+].[Na+]

Specifications

Quantity 25 g
Linear Formula Na2SeO3 · 5H2O
UN Number 2630
Formula Weight 263.01g/mol
Percent Purity ≥99% (RT)
Chemical Name or Material Sodium Selenite Pentahydrate
compliance-icons compliance-icons
Product Identifier
  • Sodium selenite pentahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H300 + H330-Fatal if swallowed or if inhaled.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'