missing translation for 'onlineSavingsMsg'
Learn More

Sodium solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

54.00 € - 114.00 €

Chemical Identifiers

CAS 7440-23-5
Molecular Formula Na
Molecular Weight (g/mol) 22.99
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES [Na]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12932195
View Documents
Fisher Chemical
A 042 100
100 mL Bottle
54.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
12942195
View Documents
Fisher Chemical
A 042 250
250 mL Bottle
77.00 €
250mL
Please to purchase this item. Need a web account? Register with us today!
12952195
View Documents
Fisher Chemical
A 042 500
500 mL Bottle
114.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

7440-23-5
22.99
KEAYESYHFKHZAL-UHFFFAOYSA-N
Na
MFCD00085307
[Na]

Specifications

1g/L
Single-component Standard Solution
MFCD00085307
[Na]
22.99
Liquid
7440-23-5
Na
KEAYESYHFKHZAL-UHFFFAOYSA-N
sodium(1+) ion
H2O
Sodium
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months. (12 Months after opening)

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.