missing translation for 'onlineSavingsMsg'
Learn More

Sodium Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™

Product Code. 11470299
Click to view available options
Quantity:
50 g
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Na1000μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 7440-23-5, 8042-47-5
Packaging Triple-leached LDPE bottle
CAS Min % 0.5, 99.5
Quantity 50 g
CAS Max % 0.5, 99.5
Certificate of Analysis With Certificate of Analysis.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Molecular Formula Na
MDL Number MFCD00085307
InChI Key KEAYESYHFKHZAL-UHFFFAOYSA-N
SMILES [Na]
IUPAC Name sodium
Molecular Weight (g/mol) 22.99
PubChem CID 5360545
ChEBI CHEBI:26708
Grade Oil/Plasma
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
Base Oil 1000μg/g ORG-NA8-2Z
Base Oil 5000μg/g ORG-NA8-4Z
Solvent or Matrix Base Oil 20
Chemical Name or Material Sodium Oil Standard
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'