missing translation for 'onlineSavingsMsg'
Learn More

Sodium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

60.80 € - 122.00 €

Specifications

Chemical Name or Material Sodium Standard
CAS 7440-23-5, 7732-18-5, 7697-37-2
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10192582
View Documents
SPEX Certiprep
PLNA2-2Y
125 mL Triple-leached LDPE bottle
60.80 €
125mL
Please to purchase this item. Need a web account? Register with us today!
10427132
View Documents
SPEX Certiprep
PLNA2-2X
500 mL Triple-leached LDPE bottle
122.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Na1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Sodium Standard
Triple-leached LDPE bottle
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
MFCD00085307
[Na]
22.99
CHEBI:26708
2% HNO3
7440-23-5, 7732-18-5, 7697-37-2
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Na
KEAYESYHFKHZAL-UHFFFAOYSA-N
sodium
5360545
Assurance
Videos
Safety and Handling

Safety and Handling

Causes severe skin burns and eye damage

missing translation for 'dotInformation' : Hazardous

Recommended Storage : Ambient

Product Identifier
  • 1000

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : nitric acid, Sodiumfrom Sodium carbonate

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.