missing translation for 'onlineSavingsMsg'
Learn More
Sodium sulfate, 99+%, ACS reagent, granular, anhydrous, Thermo Scientific Chemicals Product Code.: 13468528

Sodium sulfate, 99+%, ACS reagent, granular, anhydrous, Thermo Scientific Chemicals

Código de producto. 13468528
10 kg, Plastic drum
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 13468528

Brand: Thermo Scientific Acros 354250100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1Mostrar másMostrar menos
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Especificaciones

Boiling Point >1700.0°C
Melting Point 884.0°C
Color Colorless to Orange
pH 5.2 to 7.0 (5% soln. at 25°C)
Physical Form Liquid
Quantity 10 kg
Assay Percent Range 99+%
Linear Formula Na2SO4
Fieser 01,1103
Merck Index 15, 8809
Solubility Information Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
IUPAC Name disodium;sulfate
Formula Weight 142.04
Percent Purity ≥99%
Grade ACS Reagent
Identification Sodium : passes test (USP) Sulfate : passes test (USP)
Loss on Ignition 0.5 % max.
Packaging Plastic drum
Decomposition Information Decomposition Information: >890°C
Density 2.68g/mL
Chemical Name or Material Sodium sulfate, Anhydrous
Mostrar más Mostrar menos

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469275 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number354250025,354250010,354250100
Lot NumberA0464090 Certificate TypeCertificate of Analysis Date18/07/2024 Catalog Number354250025,354250010,354250100
Lot NumberB0155539 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number354250025,354250010,354250100
Lot NumberB0148849 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number354250025,354250010,354250100
Lot NumberB0150312 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number354250025,354250010,354250100

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Sodium sulfate, 99+%, ACS reagent, granular, anhydrous, Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado