missing translation for 'onlineSavingsMsg'
Learn More
Sodium sulfate, 99+%, for analysis, crystalline, anhydrous, Thermo Scientific Chemicals Product Code.: 10206660

Sodium sulfate, 99+%, for analysis, crystalline, anhydrous, Thermo Scientific Chemicals

Product Code. 10206660
1 kg, Plastic bottle
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10206660

Brand: Thermo Scientific Acros 219260010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium;sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Boiling Point >1700.0°C
Melting Point 884.0°C
Color White
pH 5 to 8 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula Na2SO4
Fieser 01,1103
Merck Index 15, 8809
Solubility Information Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
Formula Weight 142.04
Percent Purity ≥99%
Grade Analytical
Loss on Ignition 0.5% max.
Packaging Plastic bottle
Density 2.68g/mL
Chemical Name or Material Sodium sulfate, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465571 Certificate TypeCertificate of Analysis Date05/09/2024 Catalog Number219260010,219260100,219260025
Lot NumberA0448261 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number219260010,219260100,219260025
Lot NumberA0410956 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number219260010,219260100,219260025
Lot NumberA0437502 Certificate TypeCertificate of Analysis Date24/12/2021 Catalog Number219260010,219260100,219260025
Lot NumberA0430558 Certificate TypeCertificate of Analysis Date14/06/2021 Catalog Number219260010,219260100,219260025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium sulfate, 99+%, for analysis, crystalline, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.