missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Sodium sulfate, 99%, for biochemistry, anhydrous
65.50 € - 222.00 €
Chemical Identifiers
CAS | 7757-82-6 |
---|---|
Molecular Formula | Na2O4S |
Molecular Weight (g/mol) | 142.04 |
MDL Number | MFCD00003504 |
InChI Key | PMZURENOXWZQFD-UHFFFAOYSA-L |
Synonym | sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 |
PubChem CID | 24436 |
ChEBI | CHEBI:32149 |
IUPAC Name | disodium;sulfate |
SMILES | [Na+].[Na+].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12625127
|
Thermo Scientific Chemicals
447925000 |
500 g |
65.50 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12652936
|
Thermo Scientific Chemicals
447920025 |
2.5 kg |
222.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7757-82-6 | |
142.04 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
24436 | |
disodium;sulfate |
Na2O4S | |
MFCD00003504 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
CHEBI:32149 | |
[Na+].[Na+].[O-]S([O-])(=O)=O |
Specifications
>1700.0°C | |
7757-82-6 | |
Na2O4S | |
MFCD00003504 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium;sulfate | |
24436 | |
142.04 | |
Biochemical | |
Sodium sulfate |
884.0°C | |
99% | |
Na2SO4 | |
15,8809 | |
(10% in water) Clear colorless | |
[Na+].[Na+].[O-]S([O-])(=O)=O | |
142.04 | |
CHEBI:32149 | |
99% | |
Plastic bottle |
Safety and Handling
missing translation for 'einecsNumber' : 231-820-9
RUO – Research Use Only