missing translation for 'onlineSavingsMsg'
Learn More

Sodium sulfate, 99%, extra pure, anhydrous, Thermo Scientific Chemicals

21.31 € - 87.74 €

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10388090
View Documents Promotion Details
Thermo Scientific Acros
196640250
25 g Glass Bottle
21.31 €
25g
Please to purchase this item. Need a web account? Register with us today!
10032580
View Documents Promotion Details
Thermo Scientific Acros
196640010
1 kg Plastic bottle
27.34 €
1kg
Please to purchase this item. Need a web account? Register with us today!
10032590
View Documents Promotion Details
Thermo Scientific Acros
196640025
2.5 kg Plastic bottle
56.38 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
10493372
View Documents Promotion Details
Thermo Scientific Acros
196640050
5 kg Plastic bottle
87.74 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
[Na+].[Na+].[O-]S([O-])(=O)=O
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149

Specifications

>1700.0°C
7757-82-6
Crystalline Powder
Na2O4S
MFCD00003504
15, 8809
Solubility in water: 185g/L. Other solubilities: soluble in glycerol and hydrogen iodide, insoluble in alcohol
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
99%
2.68
884.0°C
White
98.5% min
Na2SO4
01,1103
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
142.04
Extra Pure
Sodium sulfate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-820-9

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only